Assay Details
| Gene Symbol: Fdft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC054722.1 | - | 8 | 1757 | 5694816 | 130 |
| BC138301.1 | - | 8 | 1811 | 8861320 | 130 | |
| BC138302.1 | - | 8 | 1812 | 8861321 | 130 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fdft1
Genomic Map