Assay Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025857.2 | NP_080133.1 | 1 - 2 | 113 | 71 | |
| GenBank mRNA | AK159897.1 | - | 1 - 2 | 114 | 71 | |
| BC027311.1 | - | 1 - 2 | 109 | 3964905 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aagab
Genomic Map