Assay Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180960.3 | NP_851291.1 | 1 - 2 | 182 | 66 | |
| GenBank mRNA | AU079843.1 | - | 1 - 2 | 150 | 66 | |
| X83568.1 | - | 1 - 2 | 80 | 66 | ||
| X83569.1 | - | 1 - 2 | 137 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nnat
Genomic Map