Assay Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008808.3 | NP_032834.1 | 2 - 3 | 343 | 197 | |
| GenBank mRNA | AY324648.1 | - | 2 - 3 | 332 | 197 | |
| BC003817.1 | - | 2 - 3 | 343 | 3495629 | 197 | |
| M29464.1 | - | 2 - 3 | 223 | 197 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfa
Genomic Map