Assay Details
| Gene Symbol: Rpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011292.2 | NP_035422.1 | 5 - 6 | 446 | 101 | |
| GenBank mRNA | AF260271.1 | - | 4 - 5 | 416 | 101 | |
| AK012331.1 | - | 5 - 6 | 416 | 101 | ||
| AK012445.1 | - | 5 - 6 | 416 | 101 | ||
| AK017388.1 | - | 5 - 6 | 416 | 101 | ||
| AK017427.1 | - | 5 - 6 | 416 | 101 | ||
| AK084278.1 | - | 5 - 6 | 417 | 101 | ||
| AK088166.1 | - | 5 - 6 | 416 | 101 | ||
| AK153562.1 | - | 5 - 6 | 416 | 101 | ||
| AK159980.1 | - | 5 - 6 | 416 | 101 | ||
| AK168326.1 | - | 5 - 6 | 414 | 101 | ||
| AK169217.1 | - | 5 - 6 | 414 | 101 | ||
| BC013165.1 | - | 4 - 5 | 406 | 2655358 | 101 | |
| BC081435.1 | - | 4 - 5 | 392 | 6824107 | 101 | |
| BC083166.1 | - | 4 - 5 | 397 | 6397429 | 101 | |
| BC083329.1 | - | 4 - 5 | 395 | 6824346 | 101 | |
| BC086937.1 | - | 5 - 6 | 6787671 | 101 | ||
| BC089319.1 | - | 4 - 5 | 395 | 6823996 | 101 | |
| BC093526.1 | - | 4 - 5 | 395 | 5150493 | 101 | |
| U17331.1 | - | 3 - 4 | 352 | 101 | ||
| U17332.1 | - | 3 - 4 | 352 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl9
Genomic Map