Assay Details
| Gene Symbol: Tcl1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013772.2 | NP_038800.1 | 3 - 4 | 452 | 143 | |
| GenBank mRNA | AF195490.1 | - | 3 - 4 | 454 | 143 | |
| AF195491.1 | - | 3 - 4 | 353 | 143 | ||
| BC119260.1 | - | 3 - 4 | 452 | 8734009 | 143 | |
| BC120575.1 | - | 3 - 4 | 452 | 40129498 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985732 | AF195491.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985732 | BC119260.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985732 | AF195490.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985732 | BC120575.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985732 | NM_013772.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985732 | AF195491.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985732 | BC119260.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985732 | AF195490.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985732 | BC120575.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985732 | NM_013772.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707776 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707779 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707763 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707777 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736554 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736547 | - | - |
| Whole Mouse Genome G4122A | A_51_P299954 | AF195491.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299954 | BC119260.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299954 | AF195490.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299954 | BC120575.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P299954 | NM_013772.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102629 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07736547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707779 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707780 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398107 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398108 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420570_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398105 | - | - |
| Mouse Genome 430 2.0 Array | 1420570_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1420570_PM_x_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27378_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27378_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007548.mm.1 | NM_013772.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007548.mm.1 | NM_013772.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007548.mm.1 | TC1200001057.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007549.mm.1 | NM_013772.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007549.mm.1 | NM_013772.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007549.mm.1 | TC1200001057.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007550.mm.1 | NM_013772.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007550.mm.1 | NM_013772.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007550.mm.1 | TC1200001057.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007551.mm.1 | NM_013772.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007551.mm.1 | NM_013772.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007551.mm.1 | TC1200001057.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420570_x_at | - | - |
| Mouse Expression Set 430 | 1420570_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420570_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420570_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420570_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420570_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 817692 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106040017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106040017 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2850039 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850039 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739807 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2516040 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739807 | - | - |
| mouse-6 v1.1 (Illumina) | scl0027378.2_91-S | - | - |
| mouse-6 v1.1 (Illumina) | scl42821.1.8_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850039 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106040017 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0027378.2_91-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106040017 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2921240 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739807 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739807 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2921240 | - | - |

Tcl1b3
Genomic Map