Assay Details
| Gene Symbol: Tcl1b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013776.1 | NP_038804.1 | 3 - 4 | 450 | 222 | |
| GenBank mRNA | AF195493.1 | - | 3 - 4 | 450 | 222 | |
| AK139627.1 | - | 3 - 4 | 465 | 222 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961625 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1961628 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961625 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1961628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28707686 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07736519 | - | - |
| Whole Mouse Genome G4122A | A_52_P662042 | NM_013776.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P662042 | AF195493.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P662042 | AK139627.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28707679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707677 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707664 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707695 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28707686 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10398102 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398101 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398104 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10398103 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10398100 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1200007544.mm.1 | NM_013776.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007544.mm.1 | NM_013776.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007544.mm.1 | TC1200001056.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1200007543.mm.1 | NM_013776.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007543.mm.1 | NM_013776.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200007543.mm.1 | TC1200001056.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 799854 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6510129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006510129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2744336 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2744336 | - | - |
| mouse-6 v1.1 (Illumina) | scl42822.1.16_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6510129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl42822.1.16_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2744336 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2744336 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl42822.1.16_1-S | - | - |

Tcl1b5
Genomic Map