Assay Details
| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016806.3 | NP_058086.2 | 4 - 5 | 689 | 85 | |
| NM_182650.4 | NP_872591.1 | 4 - 5 | 689 | 85 | ||
| NR_104468.1 | 4 - 5 | 689 | 85 | |||
| GenBank mRNA | AF073993.1 | - | 4 - 5 | 567 | 85 | |
| AF406651.2 | - | 3 - 4 | 478 | 85 | ||
| AK049771.1 | - | 3 - 4 | 3054 | 85 | ||
| AK089012.1 | - | 4 - 5 | 640 | 85 | ||
| BC059107.1 | - | 2 - 3 | 225 | 85 | ||
| BC141253.1 | - | 4 - 5 | 523 | 9055865 | 85 | |
| BC145588.1 | - | 4 - 5 | 523 | 9054154 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpa2b1
Genomic Map