Assay Details
| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019456.2 | NP_062329.2 | 8 - 9 | 1103 | 96 | |
| GenBank mRNA | AF020313.1 | - | 5 - 6 | 709 | 96 | |
| AK021331.1 | - | 9 - 10 | 982 | 96 | ||
| AK042467.1 | - | 8 - 9 | 1041 | 96 | ||
| BC003991.1 | - | 5 - 6 | 754 | 96 | ||
| BC023110.1 | - | 8 - 9 | 992 | 3707478 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apbb1ip
Genomic Map