Assay Details
| Gene Symbol: Wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027432.3 | NP_081708.1 | 8 - 9 | 889 | 202 | |
| GenBank mRNA | AK032897.1 | - | 6 - 7 | 428 | 202 | |
| AK048488.1 | - | 8 - 9 | 912 | 202 | ||
| AK050391.1 | - | 8 - 9 | 889 | 202 | ||
| AK078856.1 | - | 9 - 10 | 848 | 202 | ||
| AK131608.1 | - | 7 - 8 | 800 | 202 | ||
| AK163839.1 | - | 884 | 202 | |||
| AK168006.1 | - | 8 - 9 | 891 | 202 | ||
| AK168752.1 | - | 8 - 9 | 911 | 202 | ||
| AK169128.1 | - | 8 - 9 | 890 | 202 | ||
| BC005755.1 | - | 8 - 9 | 844 | 2654151 | 202 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr77
Genomic Map