Assay Details
| Gene Symbol: Olfr1312 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146362.1 | NP_666474.1 | 1 | 834 | 122 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2088348 | NM_146362.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141943 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2088348 | NM_146362.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P265792 | NM_146362.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141943 | - | - |
| Whole Mouse Genome G4122A | A_51_P265792 | NM_146362.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115605 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21527187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01734632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01174318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01563352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527229 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527185 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02262950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527204 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527208 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01442044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01734636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527195 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21527214 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10485810 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10485809 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 518558 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 380010 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239613 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239613 | - | - |
| mouse-6 v1.1 (Illumina) | scl18871.1.343_86-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18871.1.343_86-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380010 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2758099 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2758099 | - | - |

Olfr1312
Genomic Map