Assay Details
| Gene Symbol: Olfr781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146728.1 | NP_666939.1 | 1 | 363 | 114 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009871 | NM_146728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009871 | NM_146728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P273034 | NM_146728.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545562 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545531 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27545559 | - | - |
| Whole Mouse Genome G4122A | A_51_P273034 | NM_146728.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110842 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08747134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08747129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08747138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27545539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08357662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08551768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27545539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10367488 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10367487 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 896101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6420037 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420037 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2746143 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2746143 | - | - |
| mouse-6 v1.1 (Illumina) | scl00258723.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420037 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00258723.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420037 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2746142 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2819395 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2746142 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2819395 | - | - |

Olfr781
Genomic Map