Assay Details
| Gene Symbol: Otud6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152812.3 | NP_690025.2 | 3 - 4 | 534 | 125 | |
| GenBank mRNA | AK144761.1 | - | 3 - 4 | 357 | 125 | |
| AK155761.1 | - | 3 - 4 | 385 | 125 | ||
| AK166445.1 | - | 3 - 4 | 364 | 125 | ||
| AK166546.1 | - | 3 - 4 | 125 | |||
| AK169134.1 | - | 3 - 4 | 349 | 125 | ||
| BC031474.1 | - | 3 - 4 | 340 | 2599116 | 125 | |
| BC087552.1 | - | 3 - 4 | 331 | 6331952 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747172 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008106 | BC087552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2008106 | NM_152812.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646583 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 670746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112593 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39900 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160606 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130400 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008106 | BC087552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2008106 | NM_152812.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747187 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747192 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747179 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22747158 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130400 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05163230 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160606 | - | - |
| Whole Mouse Genome G4122A | A_52_P248595 | BC087552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P248595 | AK172338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P248595 | BC031474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P248595 | AK169134.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P248595 | AK172229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452302 | BC087552.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452302 | AK172338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452302 | BC006824.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452302 | BC031474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P452302 | AK169134.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115600 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130400 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705064 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03813745 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03813743 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03705054 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747169 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747170 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747172 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747173 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22747167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22746998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22747134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03813745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03302386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03602827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03476470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747192 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747194 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747116 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03617766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747181 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03537337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03949335 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747184 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747119 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22747187 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10599431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511664 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511663 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428277_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511661 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599430 | - | - |
| Mouse Genome 430 2.0 Array | 1428277_at | - | - |
| Mouse Genome 430 2.0 Array | 1428277_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72201_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72201_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400018675.mm.1 | NM_152812.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018675.mm.1 | NM_152812.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018675.mm.1 | TC0400002236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400018677.mm.1 | NM_152812.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018677.mm.1 | NM_152812.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018677.mm.1 | TC0400002236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400018672.mm.1 | NM_152812.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018672.mm.1 | NM_152812.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018672.mm.1 | TC0400002236.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400018674.mm.1 | NM_152812.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018674.mm.1 | NM_152812.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400018674.mm.1 | TC0400002236.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428277_at | - | - |
| Mouse Expression Set 430 | 1428277_PM_at | - | - |
| Murine Genome U74v2 Set | 98135_PM_r_at | - | - |
| Murine Genome U74v2 Set | 98135_r_at | - | - |
| Murine Genome U74v2 Set | 166630_PM_i_at | - | - |
| Murine Genome U74v2 Set | 166630_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428277_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428277_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428277_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428277_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1253150 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253150 | - | - |
| mouse-6 v1.1 (Illumina) | scl072201.1_290-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl072201.1_290-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1253150 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253150 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl072201.1_290-S | - | - |

Otud6b
Genomic Map