Assay Details
| Gene Symbol: Rab39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175122.6 | NP_780331.1 | 1 - 2 | 432 | 131 | |
| GenBank mRNA | AB232636.1 | - | 1 - 2 | 208 | 131 | |
| AK018257.1 | - | 1 - 2 | 417 | 131 | ||
| AK020665.1 | - | 1 - 2 | 413 | 131 | ||
| AK039013.1 | - | 1 - 2 | 406 | 131 | ||
| AK048981.1 | - | 1 - 2 | 406 | 131 | ||
| AK160737.1 | - | 1 - 2 | 406 | 131 | ||
| BC050853.1 | - | 1 - 2 | 426 | 6517891 | 131 | |
| BC052472.1 | - | 1 - 2 | 357 | 6833461 | 131 | |
| BY257784.1 | - | 1 - 2 | 432 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472232 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 502628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279267 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132770 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32472226 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09333794 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132770 | - | - |
| Whole Mouse Genome G4122A | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | BC050853.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | BC052472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK048981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK160737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P356620 | AK039013.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136065 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132770 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14580127 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14736749 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14736748 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32472232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32472193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14580117 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15075225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472217 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15144209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15144207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15167567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472169 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14844794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472185 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472168 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15144205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472170 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472179 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472149 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472175 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472193 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472171 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32472204 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605434 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453246_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435014_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605431 | - | - |
| Mouse Genome 430 2.0 Array | 1435014_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453246_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435014_at | - | - |
| Mouse Genome 430 2.0 Array | 1453246_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67790_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67790_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00015854.mm.1 | NM_175122.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015854.mm.1 | TC0X00002479.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015855.mm.1 | NM_175122.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015855.mm.1 | TC0X00002479.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00015856.mm.1 | NM_175122.6 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00015856.mm.1 | TC0X00002479.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453246_at | - | - |
| Mouse Expression Set 430 | 1435014_PM_at | - | - |
| Mouse Expression Set 430 | 1435014_at | - | - |
| Mouse Expression Set 430 | 1453246_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435014_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453246_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435014_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453246_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 381438 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6420519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239735 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239735 | - | - |
| mouse-6 v1.1 (Illumina) | scl54114.2_230-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54114.2_230-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1239735 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239735 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54114.2_230-S | - | - |

Rab39b
Genomic Map