Assay Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178069.5 | NP_835170.1 | 1 - 2 | 154 | 94 | |
| GenBank mRNA | AK030940.1 | - | 1 - 2 | 129 | 94 | |
| AK132527.1 | - | 1 - 2 | 158 | 94 | ||
| AK135345.1 | - | 149 | 94 | |||
| AK145186.1 | - | 1 - 2 | 129 | 94 | ||
| AK146895.1 | - | 1 - 2 | 129 | 94 | ||
| BC043724.1 | - | 1 - 2 | 154 | 5004563 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lsg1
Genomic Map