Assay Details
| Gene Symbol: Dsg1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181682.1 | NP_859010.1 | 3 - 4 | 311 | 146 | |
| GenBank mRNA | AK036986.1 | - | 3 - 4 | 419 | 146 | |
| AY192158.1 | - | 3 - 4 | 311 | 146 | ||
| AY315940.1 | - | 211 | 146 | |||
| BC137927.1 | - | 3 - 4 | 302 | 8860946 | 146 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115900 | BC137927.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115900 | AY192158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115900 | NM_181682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115900 | AK036986.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525547 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109619 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 483033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263603 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31124 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121581 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136359 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45501 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102424 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135096 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115900 | BC137927.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115900 | AY192158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115900 | NM_181682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115900 | AK036986.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167156 | AY315940.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167156 | BC137927.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167156 | AY192158.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167156 | NM_181682.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167156 | AK016043.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167153 | AY315940.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167153 | BC137927.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167153 | AY192158.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167153 | NM_181682.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P167153 | AK016043.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P388454 | X74335.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P388454 | BC154410.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P388454 | AK030230.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P388454 | AK029294.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P388454 | NM_010079.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539213 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539205 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31539191 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08930538 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08930522 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08930482 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135096 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102424 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08930513 | - | - |
| Whole Mouse Genome G4122A | A_51_P388454 | X74335.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388454 | BC154410.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388454 | AK030230.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388454 | AK029294.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P388454 | NM_010079.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167153 | AY315940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167153 | BC137927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167153 | AY192158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167153 | NM_181682.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167153 | AK016043.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167156 | AY315940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167156 | BC137927.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167156 | AY192158.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167156 | NM_181682.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P167156 | AK016043.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134904 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135096 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08930513 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102424 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13881390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31539249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13906663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13632293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539205 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539172 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13597682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13881402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13687787 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13797856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539215 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13881398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31539194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13881366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13620265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10454131 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454128 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454130 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10454120 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455519_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454113 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10454129 | - | - |
| Mouse Genome 430 2.0 Array | 1455519_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455519_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800001050.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001050.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001059.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001059.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001045.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001045.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001047.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001047.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001054.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001054.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001055.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001055.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001056.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001056.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001046.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001046.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001048.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001048.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001051.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001051.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001053.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001053.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001052.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001052.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001057.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001057.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001058.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001058.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001044.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001044.mm.1 | TC1800000154.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800001049.mm.1 | NM_181682.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800001049.mm.1 | TC1800000154.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455519_at | - | - |
| Mouse Expression Set 430 | 1455519_PM_at | - | - |
| Murine Genome U74v2 Set | 166420_at | - | - |
| Murine Genome U74v2 Set | 166420_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455519_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455519_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 451101 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3850204 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3360286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3850204 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003360286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850204 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2631569 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2751822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2751822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2631569 | - | - |
| mouse-6 v1.1 (Illumina) | scl0225256.19_24-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00225256.1_88-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850204 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3360286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00225256.1_88-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0225256.19_24-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3360286 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850204 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2751822 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631568 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2631569 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631568 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631569 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2751822 | - | - |

Dsg1b
Genomic Map