Assay Details
| Gene Symbol: Cox6b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289848.1 | NP_001276777.1 | 3 - 4 | 399 | 78 | |
| NM_183405.2 | NP_899664.1 | 3 - 4 | 368 | 78 | ||
| NM_183406.3 | NP_899665.1 | 2 - 3 | 279 | 78 | ||
| NR_110411.1 | 3 - 4 | 379 | 78 | |||
| GenBank mRNA | AK160152.1 | - | 3 - 4 | 298 | 78 | |
| AY152400.1 | - | 3 - 4 | 368 | 78 | ||
| BC048670.1 | - | 2 - 3 | 279 | 6773974 | 78 | |
| BC107296.1 | - | 3 - 4 | 325 | 40053107 | 78 | |
| BC107297.1 | - | 3 - 4 | 325 | 40053112 | 78 | |
| BC125562.1 | - | 3 - 4 | 308 | 40130077 | 78 | |
| BC125564.1 | - | 3 - 4 | 311 | 40130079 | 78 | |
| BC145201.1 | - | 3 - 4 | 343 | 9053752 | 78 | |
| CT010366.1 | - | 2 - 3 | 212 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796150 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168426 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950306 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950308 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950310 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950313 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950307 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24950317 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168426 | - | - |
| Whole Mouse Genome G4122A | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | AK160152.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | AY152400.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC048670.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC107296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P300506 | BC107297.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132110 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168426 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06258404 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06258397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24950310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06080894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06537409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950303 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258408 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950311 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06258397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24950304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06282928 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559652 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559656 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559654 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435275_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559649 | - | - |
| Mouse Genome 430 2.0 Array | 1435275_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435275_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 333182_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 333182_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NM_001289848.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NM_001289849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NM_001289850.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NM_183405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NM_183406.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | NR_110411.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019806.mm.1 | TC0700002169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NM_001289848.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NM_001289849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NM_001289850.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NM_183405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NM_183406.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | NR_110411.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019808.mm.1 | TC0700002169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NM_001289848.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NM_001289849.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NM_001289850.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NM_183405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NM_183406.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | NR_110411.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019804.mm.1 | TC0700002169.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019810.mm.1 | NM_001289848.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019810.mm.1 | NM_183405.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019810.mm.1 | NM_183406.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019810.mm.1 | NR_110411.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019810.mm.1 | TC0700002169.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435275_at | - | - |
| Mouse Expression Set 430 | 1435275_PM_at | - | - |
| Murine Genome U74v2 Set | 97351_i_at | - | - |
| Murine Genome U74v2 Set | 97352_f_at | - | - |
| Murine Genome U74v2 Set | 97352_PM_f_at | - | - |
| Murine Genome U74v2 Set | 160813_r_at | - | - |
| Murine Genome U74v2 Set | 97351_PM_i_at | - | - |
| Murine Genome U74v2 Set | 160813_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171262_PM_r_at | - | - |
| Murine Genome U74v2 Set | 171262_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435275_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1435275_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435275_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1435275_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 310801 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1340010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001340010 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2764551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229627 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2764551 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1229627 | - | - |
| mouse-6 v1.1 (Illumina) | scl31804.3.1_22-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00097.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120039 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1340010 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31804.3.1_22-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00097.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1340010 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120039 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1229627 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2764551 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2764551 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229627 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00097.1_9-S | - | - |

Cox6b2
Genomic Map