Assay Details
| Gene Symbol: Gab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010248.2 | NP_034378.2 | 7 - 8 | 1704 | 70 | |
| GenBank mRNA | AF104244.1 | - | 7 - 8 | 1704 | 70 | |
| BC103525.1 | - | 7 - 8 | 1679 | 40039280 | 70 | |
| BC103526.1 | - | 7 - 8 | 1679 | 40039281 | 70 | |
| BC103527.1 | - | 7 - 8 | 1679 | 40039282 | 70 | |
More Information
Set Membership: |
|

Gab2
Genomic Map