Assay Details
| Gene Symbol: Stat5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164062.1 | NP_001157534.1 | 9 - 10 | 1273 | 69 | |
| NM_011488.3 | NP_035618.1 | 10 - 11 | 1503 | 69 | ||
| GenBank mRNA | AK154070.1 | - | 9 - 10 | 1238 | 69 | |
| AY299491.1 | - | 9 - 10 | 1251 | 69 | ||
| AY299492.1 | - | 9 - 10 | 1251 | 69 | ||
| BC008998.1 | - | 9 - 10 | 1210 | 3482404 | 69 | |
| BC013274.1 | - | 10 - 11 | 1462 | 4036030 | 69 | |
| BC056647.1 | - | 9 - 10 | 1271 | 5033894 | 69 | |
| EU249543.1 | - | 9 - 10 | 1242 | 69 | ||
| U21103.1 | - | 10 - 11 | 1351 | 69 | ||
| U36502.1 | - | 10 - 11 | 1341 | 69 | ||
| Z48538.1 | - | 9 - 10 | 1263 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Stat5a
Genomic Map