Assay Details
| Gene Symbol: Tnfrsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021349.1 | NP_067324.1 | 2 - 3 | 343 | 77 | |
| GenBank mRNA | AF257673.1 | - | 2 - 3 | 343 | 77 | |
| AK004668.1 | - | 3 - 4 | 420 | 77 | ||
| BC139392.1 | - | 2 - 3 | 400 | 8862414 | 77 | |
| BC139393.1 | - | 2 - 3 | 400 | 8862415 | 77 | |
| BC141867.1 | - | 2 - 3 | 343 | 40054144 | 77 | |
| BC141868.1 | - | 2 - 3 | 343 | 40054146 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846773 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846772 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169559 | BC141867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169559 | BC141868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169559 | AK004668.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169559 | BC139392.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2169559 | BC139393.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981972 | BC141867.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981972 | BC141868.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981972 | AK155178.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981972 | BC139392.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1981972 | BC139393.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22735 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155332 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86544 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981972 | BC141867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981972 | BC141868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981972 | AK155178.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981972 | BC139392.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1981972 | BC139393.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169559 | BC141867.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169559 | BC141868.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169559 | AK004668.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169559 | BC139392.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2169559 | BC139393.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P106269 | BC141867.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P106269 | BC141868.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P106269 | AK004668.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P106269 | BC139392.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P106269 | BC139393.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02340257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02340266 | - | - |
| Whole Mouse Genome G4122A | A_51_P106269 | BC141867.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P106269 | BC141868.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P106269 | AK004668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P106269 | BC139392.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P106269 | BC139393.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100036 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129932 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846774 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846772 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27846773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27846739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09219805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09219807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09219802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09450599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09521726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09240216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09123465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09503315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09450601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09332855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09599172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27846708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10376763 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376757 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376762 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376758 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376759 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376761 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10376760 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423182_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10376755 | - | - |
| Mouse Genome 430 2.0 Array | 1423182_at | - | - |
| Mouse Genome 430 2.0 Array | 1423182_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100006201.mm.1 | NM_021349.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006201.mm.1 | NM_021349.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006201.mm.1 | TC1100000774.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100006200.mm.1 | NM_021349.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006200.mm.1 | NM_021349.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006200.mm.1 | TC1100000774.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100006202.mm.1 | NM_021349.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006202.mm.1 | NM_021349.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100006202.mm.1 | TC1100000774.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1423182_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423182_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 642510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102640397 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1990373 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102640397 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001990373 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2480814 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2422416 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2480814 | - | - |
| mouse-6 v1.1 (Illumina) | scl0057916.2_218-S | - | - |
| mouse-6 v1.1 (Illumina) | scl41469.8_427-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1990373 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102640397 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0057916.2_218-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102640397 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1990373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2480814 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2480814 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2422416 | - | - |

Tnfrsf13b
Genomic Map