Assay Details
| Gene Symbol: Ppp1r14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008889.2 | NP_032915.2 | 1 | 373 | 85 | |
| GenBank mRNA | BC082545.1 | - | 1 | 373 | 6820968 | 85 |
| BC125650.1 | - | 1 | 256 | 40130165 | 85 | |
| BC125654.1 | - | 1 | 270 | 40130169 | 85 | |
| X97490.1 | - | 1 | 436 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937064 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937065 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1984751 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109263 | AK165792.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109263 | BC082545.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109263 | CF739461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109263 | X97490.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2109263 | NM_008889.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944254 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822202 | AK005466.1 | genbank_mrna_mouse |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175672 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1984751 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109263 | AK165792.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109263 | BC082545.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109263 | CF739461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109263 | X97490.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2109263 | NM_008889.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937073 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937072 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937067 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937084 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937077 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31937078 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175672 | - | - |
| Whole Mouse Genome G4122A | A_52_P262883 | AK165792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262883 | BC082545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262883 | BC125650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262883 | BC125654.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P262883 | CF739461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133037 | AK165792.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133037 | BC082545.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133037 | CF739461.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133037 | X97490.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133037 | NM_008889.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105915 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136108 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175672 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937066 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31937064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31936995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31937091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14231359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14359394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937077 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14231352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937053 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14311530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937082 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14490804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31937045 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10528144 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461024 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461025 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461023 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461027 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10461026 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436716_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450914_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1445573_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10528143 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10461022 | - | - |
| Mouse Genome 430 2.0 Array | 1450914_at | - | - |
| Mouse Genome 430 2.0 Array | 1450914_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436716_at | - | - |
| Mouse Genome 430 2.0 Array | 1445573_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445573_at | - | - |
| Mouse Genome 430 2.0 Array | 1436716_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900001717.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001717.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001717.mm.1 | TC1900000127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900001718.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001718.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001718.mm.1 | TC1900000127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900001714.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001714.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001714.mm.1 | TC1900000127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900001720.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001720.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001720.mm.1 | TC1900000127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900001719.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001719.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001719.mm.1 | TC1900000127.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900001716.mm.1 | NM_008889.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001716.mm.1 | NM_008889.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900001716.mm.1 | TC1900000127.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436716_at | - | - |
| Mouse Expression Set 430 | 1450914_PM_at | - | - |
| Mouse Expression Set 430 | 1450914_at | - | - |
| Mouse Expression Set 430 | 1436716_PM_at | - | - |
| Mouse Expression Set 430 | 1445573_PM_at | - | - |
| Mouse Expression Set 430 | 1445573_at | - | - |
| Murine Genome U74v2 Set | 160078_PM_at | - | - |
| Murine Genome U74v2 Set | 160078_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450914_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436716_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450914_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436716_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450914_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436716_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436716_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450914_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445573_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1445573_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 899637 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3800075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101190520 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2614626 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2614626 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233175 | - | - |
| mouse-6 v1.1 (Illumina) | GI_6679402-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0018938.1_81-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101190520 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0018938.1_81-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101190520 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242221 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1233175 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0018938.1_81-S | - | - |

Ppp1r14b
Genomic Map