Assay Details
| Gene Symbol: S100a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016740.3 | NP_058020.1 | 3 | 255 | 153 | |
| GenBank mRNA | AK002490.1 | - | 3 | 257 | 153 | |
| AK003889.1 | - | 3 | 255 | 153 | ||
| AK164352.1 | - | 3 | 256 | 153 | ||
| BC021916.1 | - | 3 | 234 | 4913795 | 153 | |
| BC086903.1 | - | 3 | 254 | 6764162 | 153 | |
| U41341.1 | - | 2 | 189 | 153 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336826 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336825 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336827 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2065991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82525 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161034 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505981 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124073 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2065991 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155643 | AA207524.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155643 | AK002490.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155643 | AK003889.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155643 | NM_016740.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P155643 | AK164352.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336795 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336800 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336820 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336811 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336818 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336793 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336822 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336804 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336803 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336799 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336835 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22336813 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124073 | - | - |
| Whole Mouse Genome G4122A | A_51_P155643 | AA207524.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155643 | AK002490.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155643 | AK003889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155643 | NM_016740.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155643 | AK164352.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115650 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124073 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03147799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03176320 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02775665 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03200548 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336826 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336825 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22336827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22336791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03133309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03200549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02385210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02875047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02484732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336815 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02568477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02385207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02858444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02338790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336863 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02385201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02487428 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03179432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22336845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02487420 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10457641 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10457642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10457643 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493994 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493993 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460351_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493990 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10457640 | - | - |
| Mouse Genome 430 2.0 Array | 1460351_at | - | - |
| Mouse Genome 430 2.0 Array | 1460351_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006534.mm.1 | NM_016740.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006534.mm.1 | NM_016740.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006534.mm.1 | TC0300000852.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006536.mm.1 | NM_016740.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006536.mm.1 | NM_016740.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006536.mm.1 | TC0300000852.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300006535.mm.1 | NM_016740.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006535.mm.1 | NM_016740.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006535.mm.1 | TC0300000852.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460351_at | - | - |
| Mouse Expression Set 430 | 1460351_PM_at | - | - |
| Murine Genome U74v2 Set | 98600_PM_at | - | - |
| Murine Genome U74v2 Set | 98600_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460351_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460351_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460351_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460351_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 895331 | - | - |
| ABI Mouse Genome Survey Microarray | 904145 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2260064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260064 | - | - |
| mouse-6 v1.1 (Illumina) | scl020195.2_6-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020195.2_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2883392 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1212626 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883392 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl020195.2_6-S | - | - |

S100a11
Genomic Map