Assay Details
| Gene Symbol: Arpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026369.2 | NP_080645.2 | 2 | 352 | 167 | |
| GenBank mRNA | AK008097.1 | - | 2 | 167 | ||
| AK011348.1 | - | 2 | 229 | 167 | ||
| AK011911.1 | - | 2 | 229 | 167 | ||
| AK017669.1 | - | 2 | 230 | 167 | ||
| AK017985.1 | - | 2 | 263 | 167 | ||
| AK019149.1 | - | 2 | 229 | 167 | ||
| AK087427.1 | - | 2 | 219 | 167 | ||
| AK149756.1 | - | 2 | 221 | 167 | ||
| AK150443.1 | - | 2 | 229 | 167 | ||
| AK150703.1 | - | 2 | 229 | 167 | ||
| AK151490.1 | - | 2 | 214 | 167 | ||
| AK151654.1 | - | 2 | 220 | 167 | ||
| AK151656.1 | - | 2 | 242 | 167 | ||
| AK152281.1 | - | 2 | 242 | 167 | ||
| AK153262.1 | - | 2 | 223 | 167 | ||
| AK159407.1 | - | 2 | 276 | 167 | ||
| AK159438.1 | - | 2 | 216 | 167 | ||
| AK159738.1 | - | 2 | 239 | 167 | ||
| AK166284.1 | - | 2 | 244 | 167 | ||
| AK166335.1 | - | 2 | 259 | 167 | ||
| AK166725.1 | - | 2 | 243 | 167 | ||
| AK166919.1 | - | 2 | 245 | 167 | ||
| AK167916.1 | - | 2 | 213 | 167 | ||
| AK168264.1 | - | 2 | 220 | 167 | ||
| AK168986.1 | - | 2 | 191 | 167 | ||
| BC060143.1 | - | 2 | 258 | 6411792 | 167 | |
| CF170085.1 | - | 2 | 352 | 167 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc5
Genomic Map