Assay Details
| Gene Symbol: Rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052835.3 | NP_443067.1 | 6 - 7 | 571 | 72 | |
| GenBank mRNA | AK011022.1 | - | 6 - 7 | 531 | 72 | |
| AK012561.1 | - | 5 - 6 | 528 | 72 | ||
| AK014054.1 | - | 6 - 7 | 533 | 72 | ||
| AK088777.1 | - | 6 - 7 | 532 | 72 | ||
| AK144831.1 | - | 6 - 7 | 531 | 72 | ||
| AK146217.1 | - | 6 - 7 | 532 | 72 | ||
| AK151416.1 | - | 6 - 7 | 531 | 72 | ||
| AK151655.1 | - | 6 - 7 | 531 | 72 | ||
| AK151736.1 | - | 6 - 7 | 530 | 72 | ||
| AK152123.1 | - | 6 - 7 | 531 | 72 | ||
| AK152854.1 | - | 6 - 7 | 531 | 72 | ||
| AK153345.1 | - | 6 - 7 | 531 | 72 | ||
| AK160618.1 | - | 6 - 7 | 531 | 72 | ||
| AK166610.1 | - | 6 - 7 | 530 | 72 | ||
| AK167239.1 | - | 5 - 6 | 567 | 72 | ||
| AK167966.1 | - | 6 - 7 | 531 | 72 | ||
| AK168854.1 | - | 6 - 7 | 531 | 72 | ||
| BC006068.1 | - | 5 - 6 | 490 | 72 | ||
| BC024901.1 | - | 5 - 6 | 492 | 4224016 | 72 | |
| BC048872.1 | - | 5 - 6 | 495 | 5052784 | 72 | |
| BC082293.1 | - | 5 - 6 | 492 | 6486491 | 72 | |
| BC083327.1 | - | 5 - 6 | 492 | 5684015 | 72 | |
| BC086917.1 | - | 6 - 7 | 530 | 6768088 | 72 | |
| BC092383.1 | - | 5 - 6 | 516 | 5710462 | 72 | |
| BC098204.1 | - | 5 - 6 | 516 | 6410755 | 72 | |
| BC099437.1 | - | 5 - 6 | 495 | 30843551 | 72 | |
| M93980.1 | - | 6 - 7 | 530 | 72 | ||
| X75312.1 | - | 5 - 6 | 523 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467191 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467190 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142191 | M93980.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142191 | BC024901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142191 | BC006068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142191 | BC082293.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2142191 | BC092383.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968729 | BC099437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968729 | BC083327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968729 | BC086917.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1968729 | AK011022.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2005081 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153643 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2005081 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968729 | BC099437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968729 | BC083327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968729 | BC086917.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1968729 | AK011022.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142191 | M93980.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142191 | BC024901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142191 | BC006068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142191 | BC082293.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2142191 | BC092383.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P171216 | U89405.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495895 | M93980.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495895 | X75312.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495895 | BC099437.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495895 | BC024901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P495895 | BC048872.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467199 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467191 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467197 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467200 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467186 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467189 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467182 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467194 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467183 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32467174 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153643 | - | - |
| Whole Mouse Genome G4122A | A_52_P171216 | U89405.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495895 | M93980.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495895 | X75312.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495895 | BC099437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495895 | BC024901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P495895 | BC048872.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104663 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136695 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15154586 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15154589 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15154592 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15154609 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467189 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467191 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32467190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32467238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15154586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14740092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467191 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467188 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15154598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467158 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15214436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467174 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467145 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15154603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467156 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15154589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467154 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467184 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14659809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15154601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14518282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14781993 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14943755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32467200 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10467257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10596380 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10593322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10485655 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10600350 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448157_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10485654 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10593320 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10596379 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581505 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10600349 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10467256 | - | - |
| Mouse Genome 430 2.0 Array | 1415942_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415942_at | - | - |
| Mouse Genome 430 2.0 Array | 1448157_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1448157_s_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00004711.mm.1 | NM_052835.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004711.mm.1 | NM_052835.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004711.mm.1 | TC0X00000704.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004709.mm.1 | NM_052835.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004709.mm.1 | NM_052835.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004709.mm.1 | TC0X00000704.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004705.mm.1 | NM_052835.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004705.mm.1 | NM_052835.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004705.mm.1 | TC0X00000704.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004706.mm.1 | NM_052835.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004706.mm.1 | NM_052835.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004706.mm.1 | TC0X00000704.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00004712.mm.1 | NM_052835.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004712.mm.1 | NM_052835.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00004712.mm.1 | TC0X00000704.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1415942_PM_at | - | - |
| Mouse Expression Set 430 | 1448157_s_at | - | - |
| Mouse Expression Set 430 | 1448157_PM_s_at | - | - |
| Mouse Expression Set 430 | 1415942_at | - | - |
| Murine Genome U74v2 Set | 161657_PM_f_at | - | - |
| Murine Genome U74v2 Set | 98342_at | - | - |
| Murine Genome U74v2 Set | 161657_f_at | - | - |
| Murine Genome U74v2 Set | 98342_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415942_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448157_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415942_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448157_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 896112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940121 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940121 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940121 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2740432 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2740432 | - | - |
| mouse-6 v1.1 (Illumina) | scl0110954.5_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940121 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0110954.5_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940121 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2740432 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2740432 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0110954.5_51-S | - | - |

Rpl10
Genomic Map