Assay Details
| Gene Symbol: Ube2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016786.3 | NP_058066.2 | 1 - 2 | 285 | 122 | |
| GenBank mRNA | AB011081.1 | - | 1 - 2 | 223 | 122 | |
| AK018366.1 | - | 1 - 2 | 259 | 122 | ||
| AK036068.1 | - | 1 - 2 | 226 | 122 | ||
| AK048194.1 | - | 1 - 2 | 276 | 122 | ||
| AK139564.1 | - | 1 - 2 | 302 | 122 | ||
| AK160728.1 | - | 1 - 2 | 245 | 122 | ||
| AK162142.1 | - | 1 - 2 | 226 | 122 | ||
| AK166647.1 | - | 1 - 2 | 194 | 122 | ||
| BC002013.1 | - | 1 - 2 | 133 | 3496393 | 122 | |
| BC085311.1 | - | 1 - 2 | 211 | 30148009 | 122 | |
| BY247382.1 | - | 1 - 2 | 286 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube2k
Genomic Map