Assay Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175009.3 | NP_778174.1 | 1 - 2 | 131 | 88 | |
| GenBank mRNA | AF173297.1 | - | 1 - 2 | 151 | 88 | |
| AK032957.1 | - | 1 - 2 | 131 | 88 | ||
| AK160628.1 | - | 1 - 2 | 98 | 88 | ||
| AK162511.1 | - | 1 - 2 | 128 | 88 | ||
| BC048361.1 | - | 1 - 2 | 137 | 4012930 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF173297.1,AK032957.1,BC048361.1,NM_175009.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eny2
Genomic Map