Assay Details
| Gene Symbol: 2210018M11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172280.2 | NP_758484.2 | 1 - 2 | 91 | 65 | |
| GenBank mRNA | AK032985.1 | - | 1 - 2 | 118 | 65 | |
| AK137691.1 | - | 1 - 2 | 307 | 65 | ||
| BC039956.1 | - | 1 - 2 | 82 | 65 | ||
| BC066050.1 | - | 1 - 2 | 68 | 65 | ||
| BC089304.1 | - | 1 - 2 | 91 | 6854147 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK032985.1, BC089304.1, NM_172280.2 transcripts.
Set Membership: |
|

2210018M11Rik
Genomic Map