Assay Details
| Gene Symbol: Smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252591.2 | NP_001239520.1 | 3 - 4 | 487 | 70 | |
| NR_045569.1 | 4 - 5 | 527 | 70 | |||
| NR_103722.1 | 3 - 4 | 487 | 70 | |||
| GenBank mRNA | AF364070.1 | - | 3 - 4 | 326 | 70 | |
| AI035961.1 | - | 3 - 4 | 301 | 70 | ||
| AJ245772.1 | - | 3 - 4 | 342 | 70 | ||
| AK003105.1 | - | 3 - 4 | 334 | 70 | ||
| AK009857.1 | - | 3 - 4 | 334 | 70 | ||
| AK010172.1 | - | 3 - 4 | 334 | 70 | ||
| AY026524.1 | - | 3 - 4 | 333 | 70 | ||
| BC051197.1 | - | 3 - 4 | 286 | 1397202 | 70 | |
| BC061051.1 | - | 3 - 4 | 248 | 30300606 | 70 | |
| BC132358.1 | - | 3 - 4 | 162 | 40130635 | 70 | |
| BC138801.1 | - | 3 - 4 | 162 | 8861823 | 70 | |
| W18646.1 | - | 4 - 5 | 240 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Smpx
Genomic Map