Assay Details
| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002760.1 | - | 2 - 3 | 391 | 116 | |
| AK018331.1 | - | 3 - 4 | 392 | 116 | ||
| AK152116.1 | - | 3 - 4 | 454 | 116 | ||
| AK152378.1 | - | 3 - 4 | 405 | 116 | ||
| BC003334.1 | - | 3 - 4 | 408 | 3256725 | 116 | |
| BG084215.2 | - | 3 - 4 | 410 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stard3nl
Genomic Map