Assay Details
| Gene Symbol: Zkscan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139141.4 | NP_631880.2 | 3 - 4 | 842 | 92 | |
| GenBank mRNA | AK032718.1 | - | 3 - 4 | 798 | 92 | |
| AK134299.1 | - | 3 - 4 | 827 | 92 | ||
| BC050152.1 | - | 3 - 4 | 778 | 92 | ||
More Information
Set Membership: |
|

Zkscan8
Genomic Map