| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300009790.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009790.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009790.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009790.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009790.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009796.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009796.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009796.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009795.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009795.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009795.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009795.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009795.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009794.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009794.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009794.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009794.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009794.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009797.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009797.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009797.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009792.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009792.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009792.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009792.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009792.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009791.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009791.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009791.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009791.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009791.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009789.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009789.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009789.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009789.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009789.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009798.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009798.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009798.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009788.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009788.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009788.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009788.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009788.mm.1 |
TC1300001547.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300009793.mm.1 |
NM_001251833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009793.mm.1 |
NM_001251833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009793.mm.1 |
NM_139141.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009793.mm.1 |
NM_139141.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300009793.mm.1 |
TC1300001547.mm.1 |
MTA TC |