Assay Details
| Gene Symbol: Dennd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172477.4 | NP_766065.1 | 10 - 11 | 2329 | 85 | |
| GenBank mRNA | AK048041.1 | - | 10 - 11 | 2314 | 85 | |
| AK076686.1 | - | 2 - 3 | 472 | 85 | ||
| AK081176.1 | - | 10 - 11 | 2330 | 85 | ||
| AK135781.1 | - | 8 - 9 | 874 | 85 | ||
| AK160784.1 | - | 10 - 11 | 2091 | 85 | ||
| AK165202.1 | - | 10 - 11 | 2184 | 85 | ||
| AK167298.1 | - | 10 - 11 | 2183 | 85 | ||
| BC056367.1 | - | 10 - 11 | 2199 | 85 | ||
| BC057310.1 | - | 10 - 11 | 2199 | 85 | ||
| BC145599.1 | - | 9 - 10 | 2008 | 9054165 | 85 | |
| BC150799.1 | - | 9 - 10 | 2008 | 9087710 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2a
Genomic Map