Assay Details
| Gene Symbol: Dennd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172477.4 | NP_766065.1 | 14 - 15 | 2873 | 107 | |
| GenBank mRNA | AK048041.1 | - | 14 - 15 | 2858 | 107 | |
| AK076686.1 | - | 6 - 7 | 1017 | 107 | ||
| AK081176.1 | - | 14 - 15 | 2874 | 107 | ||
| AK135781.1 | - | 12 - 13 | 1418 | 107 | ||
| AK160784.1 | - | 14 - 15 | 2635 | 107 | ||
| AK165202.1 | - | 14 - 15 | 2728 | 107 | ||
| AK167298.1 | - | 14 - 15 | 2727 | 107 | ||
| BC056367.1 | - | 14 - 15 | 2743 | 107 | ||
| BC057310.1 | - | 14 - 15 | 2743 | 107 | ||
| BC145599.1 | - | 13 - 14 | 2552 | 9054165 | 107 | |
| BC150799.1 | - | 13 - 14 | 2552 | 9087710 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dennd2a
Genomic Map