Assay Details
| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009865.2 | NP_033995.1 | 3 - 4 | 1010 | 147 | |
| GenBank mRNA | AF183946.1 | - | 3 - 4 | 779 | 147 | |
| AK043661.1 | - | 3 - 4 | 753 | 147 | ||
| AK138606.1 | - | 3 - 4 | 1012 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh10
Genomic Map