Assay Details
| Gene Symbol: Psma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011965.2 | NP_036095.1 | 2 - 3 | 163 | 100 | |
| GenBank mRNA | AF060088.1 | - | 2 - 3 | 87 | 100 | |
| AJ272019.1 | - | 2 - 3 | 116 | 100 | ||
| AK088673.1 | - | 2 - 3 | 133 | 100 | ||
| AK135326.1 | - | 2 - 3 | 101 | 100 | ||
| AK137054.1 | - | 2 - 3 | 105 | 100 | ||
| AK144784.1 | - | 2 - 3 | 118 | 100 | ||
| AK161611.1 | - | 2 - 3 | 105 | 100 | ||
| AK162277.1 | - | 2 - 3 | 110 | 100 | ||
| AK167689.1 | - | 2 - 3 | 101 | 100 | ||
| BC005762.1 | - | 2 - 3 | 100 | 2655483 | 100 | |
| BY108158.1 | - | 2 - 3 | 163 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF060088.1, AJ272019.1, AK088673.1, AK137054.1, AK162277.1, AK167689.1, BC005762.1, NM_011965.2 transcripts.
Set Membership: |
|

Psma1
Genomic Map