Assay Details
| Gene Symbol: Klhl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028145.1 | NP_082421.1 | 4 - 5 | 1263 | 62 | |
| GenBank mRNA | AK013012.1 | - | 4 - 5 | 1241 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl35
Genomic Map