Assay Details
| Gene Symbol: Mapre2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162941.1 | NP_001156413.1 | 6 - 7 | 828 | 96 | |
| NM_001162942.1 | NP_001156414.1 | 6 - 7 | 771 | 96 | ||
| NM_153058.4 | NP_694698.3 | 5 - 6 | 879 | 96 | ||
| GenBank mRNA | AK028823.1 | - | 6 - 7 | 829 | 96 | |
| AK038562.1 | - | 5 - 6 | 866 | 96 | ||
| AK040767.1 | - | 5 - 6 | 854 | 96 | ||
| AK045493.1 | - | 5 - 6 | 762 | 96 | ||
| AK045904.1 | - | 5 - 6 | 854 | 96 | ||
| AK046095.1 | - | 5 - 6 | 822 | 96 | ||
| AK046221.1 | - | 5 - 6 | 846 | 96 | ||
| AK148954.1 | - | 5 - 6 | 854 | 96 | ||
| BC025804.1 | - | 5 - 6 | 826 | 5101879 | 96 | |
| BC027056.1 | - | 5 - 6 | 845 | 5361238 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapre2
Genomic Map