Assay Details
| Gene Symbol: Phtf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172992.3 | NP_766580.3 | 3 - 4 | 304 | 136 | |
| GenBank mRNA | AK035536.1 | - | 3 - 4 | 258 | 136 | |
| AK037021.1 | - | 3 - 4 | 191 | 136 | ||
| AK042382.1 | - | 3 - 4 | 283 | 136 | ||
| AK042790.1 | - | 3 - 4 | 226 | 136 | ||
| AK152951.1 | - | 3 - 4 | 257 | 136 | ||
| AW060751.1 | - | 3 - 4 | 136 | |||
| BC052841.1 | - | 3 - 4 | 217 | 30050876 | 136 | |
| CB245749.1 | - | 3 - 4 | 304 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phtf2
Genomic Map