Assay Details
| Gene Symbol: Sult1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133670.1 | NP_598431.1 | 1 - 2 | 56 | 61 | |
| GenBank mRNA | BC005413.1 | - | 1 - 2 | 56 | 3488459 | 61 |
| L02331.1 | - | 1 - 2 | 72 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC005413.1, NM_133670.1 transcripts.
Set Membership: |
|

Sult1a1
Genomic Map