Assay Details
| Gene Symbol: Nipal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145469.5 | NP_663444.2 | 1 - 2 | 147 | 62 | |
| GenBank mRNA | AK033975.1 | - | 1 - 2 | 148 | 62 | |
| AK038556.1 | - | 1 - 2 | 288 | 62 | ||
| AK079085.1 | - | 1 - 2 | 580 | 62 | ||
| AK087325.1 | - | 1 - 2 | 184 | 62 | ||
| AK132530.1 | - | 1 - 2 | 175 | 62 | ||
| BC016107.1 | - | 1 - 2 | 170 | 4921508 | 62 | |
| BC030399.1 | - | 1 - 2 | 664 | 4166825 | 62 | |
| BC058207.1 | - | 1 - 2 | 313 | 5361417 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipal2
Genomic Map