Assay Details
| Gene Symbol: Vrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252447.1 | NP_001239376.1 | 3 - 4 | 524 | 102 | |
| NM_027260.3 | NP_081536.2 | 3 - 4 | 524 | 102 | ||
| GenBank mRNA | AF513620.1 | - | 2 - 3 | 188 | 102 | |
| AK012664.1 | - | 3 - 4 | 216 | 102 | ||
| AK053297.1 | - | 3 - 4 | 227 | 102 | ||
| AK089825.1 | - | 3 - 4 | 512 | 102 | ||
| AK154485.1 | - | 3 - 4 | 237 | 102 | ||
| BC013520.1 | - | 2 - 3 | 198 | 4236402 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vrk2
Genomic Map