Assay Details
| Gene Symbol: Slc39a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136237.1 | NP_001129709.1 | 5 - 6 | 850 | 103 | |
| NM_028051.3 | NP_082327.2 | 4 - 5 | 936 | 103 | ||
| NM_028092.3 | NP_082368.1 | 4 - 5 | 812 | 103 | ||
| GenBank mRNA | AK007473.1 | - | 4 - 5 | 937 | 103 | |
| AK008448.1 | - | 4 - 5 | 813 | 103 | ||
| BC028990.1 | - | 5 - 6 | 811 | 5041383 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a5
Genomic Map