Assay Details
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194262.2 | NP_919238.1 | 5 - 6 | 566 | 87 | |
| NM_198122.2 | NP_937755.1 | 5 - 6 | 566 | 87 | ||
| GenBank mRNA | AK031062.1 | - | 5 - 6 | 549 | 87 | |
| AK041414.1 | - | 5 - 6 | 563 | 87 | ||
| AK136105.1 | - | 5 - 6 | 449 | 87 | ||
| AK161669.1 | - | 5 - 6 | 552 | 87 | ||
| BC137783.1 | - | 4 - 5 | 328 | 8860802 | 87 | |
More Information
Set Membership: |
|

Arid4b
Genomic Map