| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500011229.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011229.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011229.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011232.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011232.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011232.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011236.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011236.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011236.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011238.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011238.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011238.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011239.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011239.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011239.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011235.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011235.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011235.mm.1 |
TC1500001477.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500011230.mm.1 |
NR_040451.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011230.mm.1 |
NR_040451.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500011230.mm.1 |
TC1500001477.mm.1 |
MTA TC |