Assay Details
| Gene Symbol: 4930523O13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040451.1 | 6 - 7 | 524 | 103 | ||
| GenBank mRNA | AK014957.1 | - | 5 - 6 | 758 | 103 | |
| AK015877.1 | - | 6 - 7 | 526 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1700022A22 gene | 1700022A22Rik | 4933413D07Rik | -79.1kb | Anti-sense | Chr. 15 - 46324585 - 46373899 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930548G14 gene | 4930548G14Rik | 169.9kb | Anti-sense | Chr. 15 - 46623305 - 46639858 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930523O13Rik
Genomic Map