Assay Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027992.3 | NP_082268.2 | 1 - 2 | 203 | 119 | |
| GenBank mRNA | AI552875.1 | - | 1 - 2 | 219 | 119 | |
| AK009646.1 | - | 1 - 2 | 106 | 119 | ||
| AK018015.1 | - | 1 - 2 | 110 | 119 | ||
| BC050246.1 | - | 1 - 2 | 120 | 6313498 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009646.1, AK018015.1, BC050246.1, NM_027992.3 transcripts.
Set Membership: |
|

Tmem106b
Genomic Map