Assay Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027992.3 | NP_082268.2 | 2 - 3 | 309 | 137 | |
| GenBank mRNA | AK009646.1 | - | 2 - 3 | 212 | 137 | |
| AK018015.1 | - | 2 - 3 | 216 | 137 | ||
| AK148477.1 | - | 2 - 3 | 231 | 137 | ||
| BC050246.1 | - | 2 - 3 | 226 | 6313498 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009646.1, AK018015.1, BC050246.1, NM_027992.3 transcripts.
Set Membership: |
|

Tmem106b
Genomic Map