Assay Details
| Gene Symbol: Agpat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163379.1 | NP_001156851.1 | 2 - 3 | 517 | 75 | |
| NM_018862.3 | NP_061350.3 | 2 - 3 | 269 | 75 | ||
| GenBank mRNA | AK086281.1 | - | 2 - 3 | 75 | ||
| BC009651.1 | - | 2 - 3 | 269 | 3487719 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Agpat1
Genomic Map