Assay Details
| Gene Symbol: Abcb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029020.2 | NP_083296.2 | 11 - 12 | 1727 | 75 | |
| GenBank mRNA | AK014319.1 | - | 11 - 12 | 1458 | 75 | |
| AK030624.1 | - | 11 - 12 | 1454 | 75 | ||
| AK038760.1 | - | 11 - 12 | 1455 | 75 | ||
| AK049152.1 | - | 11 - 12 | 1471 | 75 | ||
| AK076315.1 | - | 11 - 12 | 1478 | 75 | ||
| AK166704.1 | - | 11 - 12 | 1461 | 75 | ||
| AK189429.1 | - | 1 - 2 | 68 | 75 | ||
| AK196341.1 | - | 1 - 2 | 133 | 75 | ||
| AK205793.1 | - | 1 - 2 | 81 | 75 | ||
| BC015301.1 | - | 11 - 12 | 1727 | 4237894 | 75 | |
| BC050242.1 | - | 1 - 2 | 442 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcb8
Genomic Map