Assay Details
| Gene Symbol: Wdr86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081441.1 | NP_001074910.1 | 5 - 6 | 1303 | 70 | |
| GenBank mRNA | BC048684.1 | - | 1 - 2 | 592 | 70 | |
| BC059069.1 | - | 4 - 5 | 3875 | 70 | ||
| CN528185.1 | - | 3 - 4 | 609 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr86
Genomic Map